accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HOONOO

CAS RN 13280-60-9

IUPAC Name: 5-amino-2-nitrobenzoic acid
Synonyms: 5-amino-2-nitrobenzoic acid
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: KZZWQCKYLNIOBT-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(=C1)C(O)=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results