accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13288-06-7

ONOOBr

CAS RN 13288-06-7

IUPAC Name: 1-(2-bromoethoxy)-4-nitrobenzene
Synonyms: 1-(2-bromoethoxy)-4-nitrobenzene
Molecular Weight (g/mol): 246.06
Molecular Formula: C8H8BrNO3
InChi Key: YQWCBDNNEZHPMA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OCCBr)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results