accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 133-07-3

IUPAC Name: 2-[(trichloromethyl)sulfanyl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: folpet
Molecular Weight (g/mol): 296.55
Molecular Formula: C9H4Cl3NO2S
InChi Key: HKIOYBQGHSTUDB-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)SN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results