accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 133-32-4

IUPAC Name: 4-(1H-indol-3-yl)butanoic acid
Synonyms: 3-indolebutyric acid caswell no. 499 kyselina 4-indol-3-ylmaselina indole 3-butyric acid indole-3-butrylate 4-indol-3-ylbutyric-acid indole 3-butyrate seradix 3-iodolebutyrate indol-3,4'-yl butyric acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: JTEDVYBZBROSJT-UHFFFAOYSA-N
SMILES: OC(=O)CCCC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (9)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (9)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)