CAS RN 1330-43-4
Thermo Scientific Chemicals Sodium borate, 0.5M buffer soln., pH 8.5
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 8.5
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Thermo Scientific Chemicals Sodium borate, 0.5M buffer soln., pH 9.0
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.5
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Sodium tetraborate, 98%, for analysis, anhydrous
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N Synonym: Borax, fused IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 8.0
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis)
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N Synonym: Borax, anhydrous; Sodium borate (tetra) IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Sodium tetraborate, anhydrous, 99.5% (metals basis)
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N Synonym: Borax, anhydrous,Sodium borate (tetra) IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Sodium tetraborate, anhydrous, 99.95% (metals basis)
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N Synonym: Borax IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Thermo Scientific Chemicals Sodium borate, 0.5M buffer soln., pH 8.0
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.0
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Borax Anhydrous MP Biomedicals
CAS: 1330-43-4 Molecular Formula: B4Na2O7 Molecular Weight (g/mol): 201.21 MDL Number: MFCD00081185 InChI Key: NXSVMNYRVBQCMO-UHFFFAOYSA-N Synonym: Sodium Tetraborate,Sodium Borate Anhydrous,Sodium Borate IUPAC Name: disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) SMILES: [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Buffer solution pH 9.180 (25°C), With fungicide, according to DIN 19266, Solstice
With fungicide, according to DIN 19266
- 1
- 2