accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OPOOCH3OCH3

CAS RN 1330-78-5

IUPAC Name: tris(4-methylphenyl) phosphate
Synonyms: tritolyl phosphate
Molecular Weight (g/mol): 368.37
Molecular Formula: C21H21O4P
InChi Key: BOSMZFBHAYFUBJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(OP(=O)(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)