CAS RN 133052-90-1
IUPAC Name:
3-{1-[3-(dimethylamino)propyl]-1H-indol-3-yl}-4-(1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms:
bim-1
Molecular Weight (g/mol):
412.49
Molecular Formula:
C25H24N4O2
InChi Key:
QMGUOJYZJKLOLH-UHFFFAOYSA-N
SMILES:
CN(C)CCCN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CNC2=CC=CC=C12
1
–
3
of
3
results
1
Bisindolylmaleimide 1, Thermo Scientific Chemicals
CAS: 133052-90-1 Molecular Formula: C25H24N4O2 Molecular Weight (g/mol): 412.493 MDL Number: MFCD00236428 InChI Key: QMGUOJYZJKLOLH-UHFFFAOYSA-N Synonym: bisindolylmaleimide i,unii-l79h6n0v6c,rbt205 inhibitor,gö 6850,bisindolylmaleimide i gf 109203x,bim-1,chembl7463,2-1-3-dimethylaminopropyl indol-3-yl-3-indol-3-yl maleimide,3-1-3-dimethylamino propyl-1h-indol-3-yl-4-1h-indol-3-yl-1h-pyrrole-2,5-dione PubChem CID: 2396 ChEBI: CHEBI:41072 IUPAC Name: 3-[1-[3-(dimethylamino)propyl]indol-3-yl]-4-(1H-indol-3-yl)pyrrole-2,5-dione SMILES: CN(C)CCCN1C=C(C2=CC=CC=C21)C3=C(C(=O)NC3=O)C4=CNC5=CC=CC=C54
3