accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FNFFF

CAS RN 133116-83-3

IUPAC Name: 2-fluoro-6-(trifluoromethyl)benzonitrile
Synonyms: 2-fluoro-6-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: OGQYJDHTHFAPRN-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results