accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3NOH

CAS RN 13325-10-5

IUPAC Name: 4-hydroxybutan-1-aminium
Synonyms: 4-hydroxybutan-1-aminium
Molecular Weight (g/mol): 90.15
Molecular Formula: C4H12NO
InChi Key: BLFRQYKZFKYQLO-UHFFFAOYSA-O
SMILES: [NH3+]CCCCO
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)