accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 133261-10-6

H3COONNH3CCH3H3CCH3

CAS RN 133261-10-6

IUPAC Name: ethyl 3-tert-butyl-1-methyl-1H-pyrazole-5-carboxylate
Synonyms: ethyl 5-tert-butyl-2-methylpyrazole-3-carboxylate
Molecular Weight (g/mol): 210.28
Molecular Formula: C11H18N2O2
InChi Key: JCOHRMJIHAFFBC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=NN1C)C(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results