accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHO

CAS RN 13331-23-2

IUPAC Name: (furan-2-yl)boronic acid
Synonyms: furan-2-ylboronic acid
Molecular Weight (g/mol): 111.89
Molecular Formula: C4H5BO3
InChi Key: PZJSZBJLOWMDRG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CO1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)