accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13333-97-6

IUPAC Name: 2-(trifluoromethyl)benzene-1-thiol
Synonyms: 2-(trifluoromethyl)benzenethiol
Molecular Weight (g/mol): 178.17
Molecular Formula: C7H5F3S
InChi Key: XVGQHRKNXSUPEF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1S
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results