CAS RN 13338-63-1
IUPAC Name:
2-(3,4,5-trimethoxyphenyl)acetonitrile
Synonyms:
2-(3,4,5-trimethoxyphenyl)acetonitrile
Molecular Weight (g/mol):
207.23
Molecular Formula:
C11H13NO3
InChi Key:
ACFJNTXCEQCDBX-UHFFFAOYSA-N
SMILES:
COC1=CC(CC#N)=CC(OC)=C1OC
1
–
1
of
1
results
1
3,4,5-Trimethoxyphenylacetonitrile, 97%
CAS: 13338-63-1 Molecular Formula: C11H13NO3 Molecular Weight (g/mol): 207.23 MDL Number: MFCD00001912 InChI Key: ACFJNTXCEQCDBX-UHFFFAOYSA-N Synonym: 3,4,5-trimethoxyphenylacetonitrile,2-3,4,5-trimethoxyphenyl acetonitrile,3,4,5-trimethoxybenzyl cyanide,3,4,5-trimethoxybenzylnitrile,3,4,5-trimethoxybenzeneacetonitrile,benzeneacetonitrile, 3,4,5-trimethoxy,3,4,5-trimethoxyphenylacetylnitryl,3,4,5-trimethoxyphenyl acetonitrile,acetonitrile, 3,4,5-trimethoxyphenyl,2-3,4,5-trimethoxyphenyl ethanenitrile PubChem CID: 25887 IUPAC Name: 2-(3,4,5-trimethoxyphenyl)acetonitrile SMILES: COC1=CC(CC#N)=CC(OC)=C1OC