accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOH3CNOO

CAS RN 13365-26-9

IUPAC Name: 1,2-dimethyl 3-nitrobenzene-1,2-dicarboxylate
Synonyms: 1,2-dimethyl 3-nitrophthalate
Molecular Weight (g/mol): 239.18
Molecular Formula: C10H9NO6
InChi Key: MLQMIKSBTAZNBK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1C(=O)OC)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results