accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 133730-34-4

H3COBOHHOOH3C

CAS RN 133730-34-4

IUPAC Name: (2,4-dimethoxyphenyl)boronic acid
Synonyms: 2,4-dimethoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: SQTUYFKNCCBFRR-UHFFFAOYSA-N
SMILES: COC1=CC=C(B(O)O)C(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results