accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NPNNClFPFFFFF

CAS RN 133894-48-1

IUPAC Name: chlorotris(pyrrolidin-1-yl)phosphanium; hexafluoro-λ⁵-phosphanuide
Synonyms: chlorotris(pyrrolidin-1-yl)phosphanium hexafluorophosphate
Molecular Weight (g/mol): 421.73
Molecular Formula: C12H24ClF6N3P2
InChi Key: BSCYRXJVGSZNKX-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.Cl[P+](N1CCCC1)(N1CCCC1)N1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results