accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNO

CAS RN 133994-99-7

IUPAC Name: 1-methyl-1H-indole-4-carbaldehyde
Synonyms: 1-methylindole-4-carbaldehyde
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: GFTNSZIYQMJJMD-UHFFFAOYSA-N
SMILES: CN1C=CC2=C(C=O)C=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results