accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNHOHNHOSOOO

CAS RN 134-31-6

IUPAC Name: bis(8-hydroxyquinolin-1-ium) sulfate
Synonyms: oxyquinoline sulfate oxyquinolinesulphate
Molecular Weight (g/mol): 388.39
Molecular Formula: C18H16N2O6S
InChi Key: YYVFXSYQSOZCOQ-UHFFFAOYSA-N
SMILES: [O-]S([O-])(=O)=O.OC1=C2[NH+]=CC=CC2=CC=C1.OC1=C2[NH+]=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results