CAS RN 13400-13-0
CAS RN 13400-13-0
Cesium fluoride, 99.9%, extra pure
CAS: 13400-13-0 Molecular Formula: CsF Molecular Weight (g/mol): 151.90 MDL Number: MFCD00010960 InChI Key: XJHCXCQVJFPJIK-UHFFFAOYSA-M Synonym: cesium fluoride,caesium fluoride,cesium monofluoride,dicesium difluoride,tricesium trifluoride,unii-t76a371hjr,caesium 1+ ion fluoride,caesiumfluoride,cesiumfluoride PubChem CID: 25953 SMILES: [F-].[Cs+]
Cesium fluoride, 99%, for analysis
CAS: 13400-13-0 Molecular Formula: CsF Molecular Weight (g/mol): 151.90 MDL Number: MFCD00010960 InChI Key: XJHCXCQVJFPJIK-UHFFFAOYSA-M Synonym: cesium fluoride,caesium fluoride,cesium monofluoride,dicesium difluoride,tricesium trifluoride,unii-t76a371hjr,caesium 1+ ion fluoride,caesiumfluoride,cesiumfluoride PubChem CID: 25953 SMILES: [F-].[Cs+]
Cesium fluoride, Puratronic™, 99.99% (metals basis)
CAS: 13400-13-0 Molecular Formula: CsF Molecular Weight (g/mol): 151.90 MDL Number: MFCD00010960 InChI Key: XJHCXCQVJFPJIK-UHFFFAOYSA-M Synonym: cesium fluoride,caesium fluoride,cesium monofluoride,dicesium difluoride,tricesium trifluoride,unii-t76a371hjr,caesium 1+ ion fluoride,caesiumfluoride,cesiumfluoride PubChem CID: 25953 IUPAC Name: cesium;fluoride SMILES: [F-].[Cs+]
Cesium fluoride, 99% (metals basis)
CAS: 13400-13-0 Molecular Formula: CsF Molecular Weight (g/mol): 151.90 MDL Number: MFCD00010960 InChI Key: XJHCXCQVJFPJIK-UHFFFAOYSA-M Synonym: cesium fluoride,caesium fluoride,cesium monofluoride,dicesium difluoride,tricesium trifluoride,unii-t76a371hjr,caesium 1+ ion fluoride,caesiumfluoride,cesiumfluoride PubChem CID: 25953 SMILES: [F-].[Cs+]
Cesium fluoride, 99.9% (metals basis)
CAS: 13400-13-0 Molecular Formula: CsF Molecular Weight (g/mol): 151.90 MDL Number: MFCD00010960 InChI Key: XJHCXCQVJFPJIK-UHFFFAOYSA-M Synonym: cesium fluoride,caesium fluoride,cesium monofluoride,dicesium difluoride,tricesium trifluoride,unii-t76a371hjr,caesium 1+ ion fluoride,caesiumfluoride,cesiumfluoride PubChem CID: 25953 SMILES: [F-].[Cs+]
Cesium fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Cesium fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Cesium fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.