accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOPOOHOHHOH2OH2OH2OH2OH2ONaNa

CAS RN 13408-09-8

IUPAC Name: [(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
Synonyms: glycerol-2-phosphate pentahydrate disodium
Molecular Weight (g/mol): 308.13
Molecular Formula: C3H19Na2O11P
InChi Key: VWVLGHQKEJESPW-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (7)
Quantity 
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (7)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (7)
  • (3)
  • (7)
  • (7)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (3)
  • (3)
  • (1)