accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13414-56-7

IUPAC Name: 2,3-dihydro-1-benzofuran-7-amine
Synonyms: 2,3-dihydro-1-benzofuran-7-amine
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: UHHZGSLXPQGPJL-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1OCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results