accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3C(S)CH3(R)OOOO

CAS RN 13429-83-9

IUPAC Name: (1R,4S)-4,7,7-trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylate
Synonyms: (1R,4S)-4,7,7-trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylate
Molecular Weight (g/mol): 197.21
Molecular Formula: C10H13O4
InChi Key: KPWKPGFLZGMMFX-ZJUUUORDSA-M
SMILES: CC1(C)[C@]2(C)CC[C@]1(OC2=O)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results