accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1343-88-0

IUPAC Name: silicon(4+) magnesium(2+) trioxidandiide
Synonyms: silicon(4+) magnesium(2+) trioxidandiide
Molecular Weight (g/mol): 100.39
Molecular Formula: MgO3Si
InChi Key: ZADYMNAVLSWLEQ-UHFFFAOYSA-N
SMILES: —
Quantity 
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (6)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)