accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2(S)OHOONH(S)HOO

CAS RN 13433-09-5

IUPAC Name: (3S)-3-azaniumyl-3-{[(1S)-1-carboxylato-2-phenylethyl]carbamoyl}propanoate
Synonyms: (3S)-3-ammonio-3-{[(1S)-1-carboxylato-2-phenylethyl]carbamoyl}propanoate
Molecular Weight (g/mol): 279.27
Molecular Formula: C13H15N2O5
InChi Key: YZQCXOFQZKCETR-UWVGGRQHSA-M
SMILES: [NH3+][C@@H](CC([O-])=O)C(=O)N[C@@H](CC1=CC=CC=C1)C([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results