accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1344-54-3

OTiOTiO

CAS RN 1344-54-3

IUPAC Name: oxo[(oxotitanio)oxy]titanium
Synonyms: titanium(III) oxide
Molecular Weight (g/mol): 143.73
Molecular Formula: O3Ti2
InChi Key: GQUJEMVIKWQAEH-UHFFFAOYSA-N
SMILES: O=[Ti]O[Ti]=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results