accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13446-03-2

MnBrBr

CAS RN 13446-03-2

IUPAC Name: manganese(2+) dibromide
Synonyms: manganese(II) bromide
Molecular Weight (g/mol): 214.75
Molecular Formula: Br2Mn
InChi Key: RJYMRRJVDRJMJW-UHFFFAOYSA-L
SMILES: [Mn++].[Br-].[Br-]
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)