accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2OMgONOOONOO

CAS RN 13446-18-9

IUPAC Name: magnesium(2+) hexahydrate dinitrate
Synonyms: magnesium(2+) hexahydrate dinitrate
Molecular Weight (g/mol): 256.4
Molecular Formula: H12MgN2O12
InChi Key: MFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (34)
  • (1)
  • (1)
Quantity 
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (9)
  • (1)
Grade 
  • (3)
  • (7)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (7)
Percent Purity 
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (10)
  • (9)
  • (2)
  • (2)
  • (34)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (9)
  • (1)
  • (3)
  • (7)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (7)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)