accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFNHOH2NO(E)

CAS RN 134470-38-5

IUPAC Name: 1-[(3E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
Synonyms: 1-[(3E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
Molecular Weight (g/mol): 316.33
Molecular Formula: C17H17FN2O3
InChi Key: UAIYNMRLUHHRMF-AATRIKPKNA-N
SMILES: CC(\C=C\C1=CC=CC(OC2=CC=C(F)C=C2)=C1)N(O)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)