accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2OH2OH2OOSOOOOSOOOOSOOOGdGd

CAS RN 13450-87-8

IUPAC Name: digadolinium(3+) octahydrate trisulfate
Synonyms: digadolinium(3+) octahydrate trisulfate
Molecular Weight (g/mol): 746.79
Molecular Formula: Gd2H16O20S3
InChi Key: JBFCVGZGWFIPNK-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Gd+3].[Gd+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results