accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13450-90-3

IUPAC Name: gallium(3+) trichloride
Synonyms: gallium trichloride
Molecular Weight (g/mol): 176.07
Molecular Formula: Cl3Ga
InChi Key: UPWPDUACHOATKO-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Ga+3]
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (4)
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (6)
Physical Form 
  • (2)
  • (3)
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (4)
  • (4)
  • (4)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)