accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

GeH4IHIHIHHI

CAS RN 13450-95-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 588.31
Molecular Formula: GeH8I4
InChi Key: WTPFKACBVKJUIS-UHFFFAOYSA-N
SMILES: [GeH4].I.I.I.I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results