accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2OTbONOOONOOONOO

CAS RN 13451-19-9

IUPAC Name: terbium(3+) hexahydrate trinitrate
Synonyms: terbium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 453.03
Molecular Formula: H12N3O15Tb
InChi Key: UTCARTSNNKGRTD-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results