CAS RN 13453-07-1
CAS RN 13453-07-1
IUPAC Name:
gold hydrochloridyl
Synonyms:
gold(III) chloride
Molecular Weight (g/mol):
232.42
Molecular Formula:
AuCl
InChi Key:
BRSVJNYNWNMJKC-UHFFFAOYSA-N
SMILES:
[Cl].[Au]
1
–
4
of
4
results
1
Gold(III) chloride, 99%
CAS: 13453-07-1 Molecular Formula: AuCl3 Molecular Weight (g/mol): 303.32 MDL Number: MFCD00014172
2
Gold(III) chloride, Au 64.4% min
CAS: 13453-07-1 Molecular Formula: AuCl3 MDL Number: MFCD00014172
3
Gold(III) chloride, Premion™, 99.99% (metals basis), Au 64.4% min
CAS: 13453-07-1 Molecular Formula: AuCl3 MDL Number: MFCD00014172
4
Gold(III) chloride, 99.99+%, (trace metal basis)
CAS: 13453-07-1 Molecular Formula: AuCl3 Molecular Weight (g/mol): 303.32