accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OOClOOOOClOOOPb

CAS RN 13453-62-8

IUPAC Name: λ²-lead(2+) trihydrate diperchlorate
Synonyms: λ²-lead(2+) trihydrate diperchlorate
Molecular Weight (g/mol): 460.1
Molecular Formula: Cl2H6O11Pb
InChi Key: KKGGEAQRICYXNM-UHFFFAOYSA-L
SMILES: O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)