accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

LiH2OH2OH2OOClOOO

CAS RN 13453-78-6

IUPAC Name: lithium(1+) trihydrate perchlorate
Synonyms: lithium perchlorate
Molecular Weight (g/mol): 160.43
Molecular Formula: ClH6LiO7
InChi Key: JAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES: [Li+].O.O.O.[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (8)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (8)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (5)
  • (3)