accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13454-74-5

OOOOOOOOOCeCeWW

CAS RN 13454-74-5

IUPAC Name: dicerium(3+) ditungsten nonaoxidandiide
Synonyms: dicerium(3+) ditungsten nonaoxidandiide
Molecular Weight (g/mol): 791.91
Molecular Formula: Ce2O9W2
InChi Key: PMMOMAJHUNGVIB-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Ce+3].[Ce+3].[W].[W]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results