accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13454-89-2

OIOOOIOOCu

CAS RN 13454-89-2

IUPAC Name: copper(2+) diiodate
Synonyms: copper(2+) diiodate
Molecular Weight (g/mol): 413.35
Molecular Formula: CuI2O6
InChi Key: LLVVIWYEOKVOFV-UHFFFAOYSA-L
SMILES: [Cu++].[O-][I](=O)=O.[O-][I](=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results