accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13455-12-4

BrBrPt

CAS RN 13455-12-4

IUPAC Name: platinum(2+) dibromide
Synonyms: platinum(II) bromide
Molecular Weight (g/mol): 354.89
Molecular Formula: Br2Pt
InChi Key: KGRJUMGAEQQVFK-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Pt++]
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)