accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1345858-76-5

CH3NFFH3CONNOCH3SCH3(?)(E/Z)(E/Z)

CAS RN 1345858-76-5

IUPAC Name: 5-{[1-(2,4-difluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]methylidene}-2-[(2-methoxyethyl)imino]-3-methyl-1,3-thiazolidin-4-one
Synonyms: 5-{[1-(2,4-difluorophenyl)-2,5-dimethylpyrrol-3-yl]methylidene}-2-[(2-methoxyethyl)imino]-3-methyl-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 405.46
Molecular Formula: C20H21F2N3O2S
InChi Key: BKQZKTRCUAWRHT-UHFFFAOYSA-N
SMILES: COCCN=C1SC(=CC2=C(C)N(C(C)=C2)C2=C(F)C=C(F)C=C2)C(=O)N1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results