accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 134615-24-0

H3C(R)OHBr

CAS RN 134615-24-0

IUPAC Name: (1R)-1-(3-bromophenyl)ethan-1-ol
Synonyms: (1R)-1-(3-bromophenyl)ethanol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: ULMJQMDYAOJNCC-ZCFIWIBFSA-N
SMILES: C[C@@H](O)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results