CAS RN 13463-40-6
IUPAC Name:
pentakis(methanidylidyneoxidanium) iron
Synonyms:
iron pentacarbonyl
Molecular Weight (g/mol):
195.9
Molecular Formula:
C5FeO5
InChi Key:
FYOFOKCECDGJBF-UHFFFAOYSA-N
SMILES:
[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
1
–
3
of
3
results
2
Iron(0) pentacarbonyl
CAS: 13463-40-6 Molecular Formula: C5FeO5 Molecular Weight (g/mol): 195.90 MDL Number: MFCD00011001 InChI Key: FYOFOKCECDGJBF-UHFFFAOYSA-N Synonym: Pentacarbonyl iron IUPAC Name: pentakis(methanidylidyneoxidanium) iron SMILES: [Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
3
Pentacarbonyliron, 99.5%
CAS: 13463-40-6 Molecular Formula: C5FeO5 Molecular Weight (g/mol): 195.90 MDL Number: MFCD00011001 InChI Key: FYOFOKCECDGJBF-UHFFFAOYSA-N Synonym: Iron pentacarbonyl; Iron carbonyl IUPAC Name: pentakis(methanidylidyneoxidanium) iron SMILES: [Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]