accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OTiO

CAS RN 13463-67-7

IUPAC Name: dioxotitanium
Synonyms: titanium dioxide titania 二氧化钛 dioxyde de titane titanium white titandioxid dioxido de titanio oxido de titanio(IV) TiO2 titanium oxide
Molecular Weight (g/mol): 79.87
Molecular Formula: O2Ti
InChi Key: GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES: O=[Ti]=O
Quantity 
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (19)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
Grade 
  • (1)
  • (2)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (19)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (4)