CAS RN 13465-09-3
IUPAC Name:
indium(3+) tribromide
Synonyms:
indium(III) bromide
Molecular Weight (g/mol):
354.53
Molecular Formula:
Br3In
InChi Key:
JKNHZOAONLKYQL-UHFFFAOYSA-K
SMILES:
[Br-].[Br-].[Br-].[In+3]
1
–
2
of
2
results
1
Indium(III) bromide, anhydrous, 99.99% (metals basis)
CAS: 13465-09-3 MDL Number: MFCD00011057
2
Indium(III) bromide, 99.99%, (trace metal basis)
CAS: 13465-09-3 Molecular Formula: Br3In Molecular Weight (g/mol): 354.52 MDL Number: MFCD00011057