accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13465-93-5

AgWO24

CAS RN 13465-93-5

IUPAC Name: tungsten(6+) disilver(1+) tetraoxidandiide
Synonyms: tungsten(6+) disilver(1+) tetraoxidandiide
Molecular Weight (g/mol): 463.57
Molecular Formula: Ag2O4W
InChi Key: JFUVYDSGHRJJDO-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Ag+].[Ag+].[W+6]
Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results