CAS RN 1346598-25-1
CAS RN 1346598-25-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Hexyl-d13-3-(1-naphthoyl)indole JWH 19-d13, TRC
CAS: 1346598-25-1 Molecular Formula: C25H12D13NO Molecular Weight (g/mol): 368.55 Synonym: (1-Hexyl-d13-1H-indol-3-yl)-1-naphthalenylmethanone SMILES: O=C(C1=CC=CC2=C1C=CC=C2)C3=CN(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C4=CC=CC=C43