accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13469-98-2

BrBrBrY

CAS RN 13469-98-2

IUPAC Name: yttrium(3+) tribromide
Synonyms: yttrium(III) bromide
Molecular Weight (g/mol): 328.62
Molecular Formula: Br3Y
InChi Key: FSDCGXUNLWDJNL-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Y+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results