accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13470-08-1

FFFTi

CAS RN 13470-08-1

IUPAC Name: titanium(3+) trifluoride
Synonyms: titanium(III) fluoride
Molecular Weight (g/mol): 104.86
Molecular Formula: F3Ti
InChi Key: NLPMQGKZYAYAFE-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Ti+3]
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)