CAS RN 134701-20-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Irganox 1141 (>85%), TRC
CAS: 134701-20-5 Molecular Formula: C24H42O Molecular Weight (g/mol): 346.59 Synonym: 2-(Hexadecan-2-yl)-4,6-dimethylphenol;2,4-Dimethyl-6-(1-methylpentadecyl)phenol IUPAC Name: 2-hexadecan-2-yl-4,6-dimethylphenol SMILES: CCCCCCCCCCCCCCC(C)c1cc(C)cc(C)c1O