accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CaOClOOOOClOOO

CAS RN 13477-36-6

IUPAC Name: calcium diperchlorate
Synonyms: calcium perchlorate
Molecular Weight (g/mol): 238.97
Molecular Formula: CaCl2O8
InChi Key: ZQAOXERLHGRFIC-UHFFFAOYSA-L
SMILES: [Ca++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results