accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH2FeOH2OH2H2OClCl

CAS RN 13478-10-9

IUPAC Name: tetrahydroxyironbis(ylium) dichloride
Synonyms: iron(II) chloride
Molecular Weight (g/mol): 198.81
Molecular Formula: Cl2FeH8O4
InChi Key: WSSMOXHYUFMBLS-UHFFFAOYSA-L
SMILES: O.O.O.O.[Cl-].[Cl-].[Fe++]
Molecular Weight (g/mol) 
  • (9)
  • (7)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (3)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (4)
  • (1)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (9)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)